Anti-IDH3A Mouse Monoclonal Antibody [clone: OTI5D2]

Proveedor: OriGene
ORIGTA500736EA 603 EUR
ORIGTA500736
Anti-IDH3A Mouse Monoclonal Antibody [clone: OTI5D2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: IDH3A (isocitrate dehydrogenase 3 (NAD+) alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR