Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI10A12]

Proveedor: OriGene
ORIGTA500706EA 603 EUR
ORIGTA500706
Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI10A12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:100

Type: Primary
Antigen: RNF144B (ring finger protein 144B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR