Anti-PSMA7 Mouse Monoclonal Antibody [clone: OTI3F11]

Proveedor: OriGene
ORIGTA500691EA 603 EUR
ORIGTA500691
Anti-PSMA7 Mouse Monoclonal Antibody [clone: OTI3F11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMA7 (proteasome (prosome, macropain) subunit, alpha type, 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR