Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI5D8]

Proveedor: OriGene
ORIGTA500680EA 603 EUR
ORIGTA500680
Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI5D8]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR