Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI2C5]

Proveedor: OriGene
ORIGTA500662EA 581 EUR
ORIGTA500662
Anti-LTA4H Mouse Monoclonal Antibody [clone: OTI2C5]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4μg/mg

Type: Primary
Antigen: LTA4H (leukotriene A4 hydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR