Anti-HSPA9 Mouse Monoclonal Antibody [clone: OTI10D7]

Proveedor: OriGene
ORIGTA500630EA 603 EUR
ORIGTA500630
Anti-HSPA9 Mouse Monoclonal Antibody [clone: OTI10D7]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HSPA9 (heat shock 70kDa protein 9 (mortalin))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR