Anti-PCSK7 Mouse Monoclonal Antibody [clone: OTI1B8]

Proveedor: OriGene
ORIGTA500628EA 608 EUR
ORIGTA500628
Anti-PCSK7 Mouse Monoclonal Antibody [clone: OTI1B8]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PCSK7 (proprotein convertase subtilisin/kexin type 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR