Anti-ALDH2 Mouse Monoclonal Antibody [clone: OTI4H2]

Proveedor: OriGene
ORIGTA500615EA 608 EUR
ORIGTA500615
Anti-ALDH2 Mouse Monoclonal Antibody [clone: OTI4H2]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: ALDH2 (aldehyde dehydrogenase 2 family (mitochondrial))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR