Anti-POR Mouse Monoclonal Antibody [clone: OTI4G2]

Proveedor: OriGene
ORIGTA500591EA 603 EUR
ORIGTA500591
Anti-POR Mouse Monoclonal Antibody [clone: OTI4G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: POR (P450 (cytochrome) oxidoreductase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR