Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI1G1]

Proveedor: OriGene
ORIGTA500590EA 603 EUR
ORIGTA500590
Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI1G1]
Anticuerpos
  • Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2μg/500μl

Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR