Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI6D11]

Proveedor: OriGene
ORIGTA500583EA 608 EUR
ORIGTA500583
Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI6D11]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2μg/500μl

Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR