Anti-PPP5C Mouse Monoclonal Antibody [clone: OTI4E1]

Proveedor: OriGene
ORIGTA500571EA 603 EUR
ORIGTA500571
Anti-PPP5C Mouse Monoclonal Antibody [clone: OTI4E1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: PPP5C (protein phosphatase 5, catalytic subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR