Anti-PRKY Mouse Monoclonal Antibody [clone: OTI2E9]

Proveedor: OriGene
ORIGTA500561EA 603 EUR
ORIGTA500561
Anti-PRKY Mouse Monoclonal Antibody [clone: OTI2E9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: PRKY (protein kinase, Y-linked, pseudogene)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR