Anti-NTRK3 Mouse Monoclonal Antibody [clone: OTI4E10]
Proveedor: OriGene
ORIGTA500538EA 603 EUR
ORIGTA500538
Anti-NTRK3 Mouse Monoclonal Antibody [clone: OTI4E10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50 Immunofluorescence: 1:100
Type: Primary
Antigen: NTRK3 (neurotrophic tyrosine kinase, receptor, type 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human