Anti-NAT8 Mouse Monoclonal Antibody [clone: OTI6H3]

Proveedor: OriGene
ORIGTA500530EA 608 EUR
ORIGTA500530
Anti-NAT8 Mouse Monoclonal Antibody [clone: OTI6H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: NAT8 (N-acetyltransferase 8 (GCN5-related, putative))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR