Anti-STAT1 Mouse Monoclonal Antibody [clone: OTI9B12]

Proveedor: OriGene
ORIGTA500521EA 603 EUR
ORIGTA500521
Anti-STAT1 Mouse Monoclonal Antibody [clone: OTI9B12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000

Type: Primary
Antigen: STAT1 (signal transducer and activator of transcription 1, 91kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR