Anti-LDHA Mouse Monoclonal Antibody [clone: OTI2C11]

Proveedor: OriGene
ORIGTA500518EA 603 EUR
ORIGTA500518
Anti-LDHA Mouse Monoclonal Antibody [clone: OTI2C11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: LDHA (lactate dehydrogenase A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR