Anti-FERMT2 Mouse Monoclonal Antibody [clone: OTI9E4]

Proveedor: OriGene
ORIGTA500504EA 608 EUR
ORIGTA500504
Anti-FERMT2 Mouse Monoclonal Antibody [clone: OTI9E4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunofluorescence: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: FERMT2 (fermitin family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR