Anti-CD4 Mouse Monoclonal Antibody [clone: OTI3B4]

Proveedor: OriGene
ORIGTA500482EA 581 EUR
ORIGTA500482
Anti-CD4 Mouse Monoclonal Antibody [clone: OTI3B4]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR