Anti-CD4 Mouse Monoclonal Antibody [clone: OTI10B5]

Proveedor: OriGene
ORIGTA500477EA 564 EUR
ORIGTA500477
Anti-CD4 Mouse Monoclonal Antibody [clone: OTI10B5]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR