Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI1C11]

Proveedor: OriGene
ORIGTA500474EA 603 EUR
ORIGTA500474
Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI1C11]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunoprecipitation: 2-4μg/mg

Type: Primary
Antigen: MAPK1 (mitogen-activated protein kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR