Anti-SERPINC1 Mouse Monoclonal Antibody [clone: OTI8D5]

Proveedor: OriGene
ORIGTA500455EA 608 EUR
ORIGTA500455
Anti-SERPINC1 Mouse Monoclonal Antibody [clone: OTI8D5]
Anticuerpos
Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINC1 (serpin peptidase inhibitor, clade C (antithrombin), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR