Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI4F2]

Proveedor: OriGene
ORIGTA500450EA 603 EUR
ORIGTA500450
Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI4F2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1 (nuclear receptor binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR