Anti-RAD9A Mouse Monoclonal Antibody [clone: OTI5D9]

Proveedor: OriGene
ORIGTA500435EA 586 EUR
ORIGTA500435
Anti-RAD9A Mouse Monoclonal Antibody [clone: OTI5D9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: RAD9A (RAD9 homolog A (S. pombe))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR