Anti-BRAF Mouse Monoclonal Antibody [clone: OTI4A5]

Proveedor: OriGene
ORIGTA500429EA 603 EUR
ORIGTA500429
Anti-BRAF Mouse Monoclonal Antibody [clone: OTI4A5]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR