Anti-SSB Mouse Monoclonal Antibody [clone: OTI6G7]

Proveedor: OriGene
TA500427
ORIGTA500427EA 616 EUR
ORIGTA500427
Anti-SSB Mouse Monoclonal Antibody [clone: OTI6G7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SSB (Sjogren syndrome antigen B (autoantigen La))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR