Anti-TYRO3 Mouse Monoclonal Antibody [clone: OTI2C4]

Proveedor: OriGene
TA500414
ORIGTA500414EA 616 EUR
ORIGTA500414
Anti-TYRO3 Mouse Monoclonal Antibody [clone: OTI2C4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: TYRO3 (TYRO3 protein tyrosine kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR