Anti-OXSR1 Mouse Monoclonal Antibody [clone: OTI1F3]

Proveedor: OriGene
ORIGTA500392EA 586 EUR
ORIGTA500392
Anti-OXSR1 Mouse Monoclonal Antibody [clone: OTI1F3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: OXSR1 (oxidative stress responsive 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR