Anti-MX1 Mouse Monoclonal Antibody [clone: OTI2G12]

Proveedor: OriGene
ORIGTA500357EA 609 EUR
ORIGTA500357
Anti-MX1 Mouse Monoclonal Antibody [clone: OTI2G12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MX1 (myxovirus (influenza virus) resistance 1, interferon-inducible protein p78 (mouse))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR