Anti-PROM2 Mouse Monoclonal Antibody [clone: OTI15F4]

Proveedor: OriGene
ORIGTA500356EA 581 EUR
ORIGTA500356
Anti-PROM2 Mouse Monoclonal Antibody [clone: OTI15F4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PROM2 (prominin 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR