Anti-NEUROG1 Mouse Monoclonal Antibody [clone: OTI5A10]

Proveedor: OriGene
ORIGTA500355EA 603 EUR
ORIGTA500355
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: OTI5A10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: NEUROG1 (neurogenin 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR