Anti-GFAP Mouse Monoclonal Antibody [clone: OTI1E11]
Proveedor: OriGene
ORIGTA500343EA 581 EUR
ORIGTA500343
Anti-GFAP Mouse Monoclonal Antibody [clone: OTI1E11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:5000; Immunofluorescence: 1:100
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human