Anti-AK1 Mouse Monoclonal Antibody [clone: OTI19G4]

Proveedor: OriGene
ORIGTA500320EA 581 EUR
ORIGTA500320
Anti-AK1 Mouse Monoclonal Antibody [clone: OTI19G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: AK1 (adenylate kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR