Anti-JUN Mouse Monoclonal Antibody [clone: OTI1C8]

Proveedor: OriGene
ORIGTA500226EA 603 EUR
ORIGTA500226
Anti-JUN Mouse Monoclonal Antibody [clone: OTI1C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR