Anti-DDK Tag Monoclonal Antibody [clone: OTI4C5]

Proveedor: OriGene
ORIGTA50011-5EA 3360 EUR
ORIGTA50011-5
Anti-DDK Tag Monoclonal Antibody [clone: OTI4C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunoprecipitation: 1:200; Immunofluorescence: 1:1000; Flow Cytometry: 1:1000

Type: Tag
Antigen: DDK Tag
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR