Anti-GATA4 Mouse Monoclonal Antibody [clone: OTI9F9]

Proveedor: OriGene
ORIGTA500091EA 608 EUR
ORIGTA500091
Anti-GATA4 Mouse Monoclonal Antibody [clone: OTI9F9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: GATA4 (GATA binding protein 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR