Anti-GSC Mouse Antibody [clone: OTI4F11]

Proveedor: OriGene
ORIGTA500087EA 581 EUR
ORIGTA500087
Anti-GSC Mouse Antibody [clone: OTI4F11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: GSC (goosecoid homeobox)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR