Anti-TNNI3 Mouse Monoclonal Antibody [clone: OTI4E5]

Proveedor: OriGene
ORIGTA500057EA 608 EUR
ORIGTA500057
Anti-TNNI3 Mouse Monoclonal Antibody [clone: OTI4E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2500-5000; Immunofluorescence: 1:50-100

Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR