Anti-FOXA1 Mouse Monoclonal Antibody [clone: OTI3C1]
Proveedor: OriGene
ORIGTA500052EA 603 EUR
ORIGTA500052
Anti-FOXA1 Mouse Monoclonal Antibody [clone: OTI3C1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000
Type: Primary
Antigen: FOXA1 (forkhead box A1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now