Anti-SOX17 Mouse Monoclonal Antibody [clone: OTI2F9]

Proveedor: OriGene
ORIGTA500045EA 581 EUR
ORIGTA500045
Anti-SOX17 Mouse Monoclonal Antibody [clone: OTI2F9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: SOX17 (SRY (sex determining region Y)-box 17)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR