Anti-SHH Mouse Monoclonal Antibody [clone: OTI10H6]

Proveedor: OriGene
ORIGTA500041EA 608 EUR
ORIGTA500041
Anti-SHH Mouse Monoclonal Antibody [clone: OTI10H6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SHH (sonic hedgehog)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR