Anti-NTF4 Mouse Monoclonal Antibody [clone: OTI4D2]

Proveedor: OriGene
ORIGTA500033EA 603 EUR
ORIGTA500033
Anti-NTF4 Mouse Monoclonal Antibody [clone: OTI4D2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:20000

Type: Primary
Antigen: NTF4 (neurotrophin 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR