Anti-KRT18 Mouse Monoclonal Antibody [clone: OTI5E8]

Proveedor: OriGene
ORIGTA500016EA 603 EUR
ORIGTA500016
Anti-KRT18 Mouse Monoclonal Antibody [clone: OTI5E8]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR