Anti-KRT18 Mouse Monoclonal Antibody [clone: OTI1E1]

Proveedor: OriGene
ORIGTA500015EA 608 EUR
ORIGTA500015
Anti-KRT18 Mouse Monoclonal Antibody [clone: OTI1E1]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR