Anti-STAT5A Mouse Monoclonal Antibody [clone: OTI4H1]

Proveedor: OriGene
ORIGTA500006EA 603 EUR
ORIGTA500006
Anti-STAT5A Mouse Monoclonal Antibody [clone: OTI4H1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:50

Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR