Anti-GDF15 Rat Monoclonal Antibody [clone: 6D12.H10.E4]

Proveedor: OriGene
TA318812
ORIGTA318812EA 680 EUR
ORIGTA318812
Anti-GDF15 Rat Monoclonal Antibody [clone: 6D12.H10.E4]
Anticuerpos
Recommended Dilutions: Immunochemistry: 10 µg/ml; Western Blot: 1:1000

Type: Primary
Antigen: GDF15 (growth differentiation factor 15)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR