Anti-THY1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA318808EA 666 EUR
ORIGTA318808
Anti-THY1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Immunochemistry: 3.75 µg/ml; Western Blot: 0.1-0.3 µg/ml

Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR