Anti-GRIN1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA318695EA 666 EUR
ORIGTA318695
Anti-GRIN1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:20000; Immunochemistry: 10 µg/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: GRIN1 (glutamate receptor, ionotropic, N-methyl D-aspartate 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR