Anti-PYGM Mouse Polyclonal Antibody [clone: 2C4]

Proveedor: OriGene
ORIGTA318625EA 867 EUR
ORIGTA318625
Anti-PYGM Mouse Polyclonal Antibody [clone: 2C4]
Anticuerpos
Recommended Dilutions: Immunochemistry: 5 µg/ml;

Type: Primary
Antigen: PYGM (phosphorylase, glycogen, muscle)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR