Anti-DOK3 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA318580EA 680 EUR
ORIGTA318580
Anti-DOK3 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:64000; Immunochemistry: 6.12 µg/ml; Western Blot: 0.1-0.3 µg/ml

Type: Primary
Antigen: DOK3 (docking protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR